CAS RN 147253-67-6
IUPAC Name:
1-[2-(2,5-dimethylphospholan-1-yl)phenyl]-2,5-dimethylphospholane
Synonyms:
1-[2-(2,5-dimethylphospholan-1-yl)phenyl]-2,5-dimethylphospholane
Molecular Weight (g/mol):
306.37
Molecular Formula:
C18H28P2
InChi Key:
AJNZWRKTWQLAJK-UHFFFAOYNA-N
SMILES:
CC1CCC(C)P1C1=CC=CC=C1P1C(C)CCC1C
1
–
1
of
1
results
1
Promotions Available
1,2-Bis[(2R,5R)-2,5-dimethyl-1-phospholanyl]benzene, 97+%
1,2-Bis[(2R,5R)-2,5-dimethyl-1-phospholanyl]benzene, 97+%
CAS: 147253-67-6 Molecular Formula: C18H28P2 Molecular Weight (g/mol): 306.37 MDL Number: MFCD00142336 InChI Key: AJNZWRKTWQLAJK-KLHDSHLOSA-N Synonym: r,r-me-duphos,methyl-duphos, r,r,r,r-methyl-duphos,unii-h5w03d1haq,h5w03d1haq,--1,2-bis 2r,5r-2,5-dimethylphospholano benzene,r,r-meduphos,methyl-duphos,-,2r,5r-me-duphos PubChem CID: 2734551 IUPAC Name: (2R,5R)-1-[2-[(2R,5R)-2,5-dimethylphospholan-1-yl]phenyl]-2,5-dimethylphospholane SMILES: CC1CCC(P1C2=CC=CC=C2P3C(CCC3C)C)C