CAS RN 147149-98-2
IUPAC Name:
2-(trifluoromethyl)pyridin-4-amine
Synonyms:
2-(trifluoromethyl)pyridin-4-amine
Molecular Weight (g/mol):
162.12
Molecular Formula:
C6H5F3N2
InChi Key:
LYNBZRJTRHTSKI-UHFFFAOYSA-N
SMILES:
NC1=CC(=NC=C1)C(F)(F)F
1
–
1
of
1
results
1
4-Amino-2-(trifluoromethyl)pyridine 98.0+%, TCI America™
CAS: 147149-98-2 Molecular Formula: C6H5F3N2 Molecular Weight (g/mol): 162.115 MDL Number: MFCD09056877 InChI Key: LYNBZRJTRHTSKI-UHFFFAOYSA-N PubChem CID: 10261388 IUPAC Name: 2-(trifluoromethyl)pyridin-4-amine SMILES: C1=CN=C(C=C1N)C(F)(F)F