accessibility menu, dialog, popup

UserName

CH3NHNOOCH3H3CCH3

CAS RN 147081-29-6

IUPAC Name: tert-butyl 3-methylpiperazine-1-carboxylate
Synonyms: tert-butyl 3-methylpiperazine-1-carboxylate
Molecular Weight (g/mol): 200.28
Molecular Formula: C10H20N2O2
InChi Key: FMLPQHJYUZTHQS-UHFFFAOYNA-N
SMILES: CC1CN(CCN1)C(=O)OC(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results