accessibility menu, dialog, popup

UserName

CAS RN 14692-17-2

OClOOOOClOOOOClOOODy

CAS RN 14692-17-2

IUPAC Name: dysprosium(3+) triperchlorate
Synonyms: dysprosium(3+) triperchlorate
Molecular Weight (g/mol): 460.84
Molecular Formula: Cl3DyO12
InChi Key: QNDHOIIJUAAORW-UHFFFAOYSA-K
SMILES: [Dy+3].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results