accessibility menu, dialog, popup

UserName

CAS RN 14591-90-3

BrBrNNPd

CAS RN 14591-90-3

IUPAC Name: palladium(2+) diamine dibromide
Synonyms: palladium(2+) diamine dibromide
Molecular Weight (g/mol): 300.29
Molecular Formula: Br2H6N2Pd
InChi Key: WLAAECNKUYLOJV-UHFFFAOYSA-L
SMILES: N.N.[Br-].[Br-].[Pd++]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results