accessibility menu, dialog, popup

UserName

CH3NH3CH3CH3CHFHFHFHFF

CAS RN 145826-81-9

IUPAC Name: tetraethylazanium tetrahydrofluoride fluoride
Synonyms: tetraethylammonium tetrahydrofluoride fluoride
Molecular Weight (g/mol): 229.28
Molecular Formula: C8H24F5N
InChi Key: ANODKKTYHRYRDA-UHFFFAOYSA-M
SMILES: F.F.F.F.[F-].CC[N+](CC)(CC)CC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results