CAS RN 14573-97-8
CAS RN 14573-97-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
{2-[3-(Dimethylamino)propoxy]phenyl}methanol, 97%, Thermo Scientific™
CAS: 14573-97-8 Molecular Formula: C12H19NO2 Molecular Weight (g/mol): 209.29 MDL Number: MFCD09064995 InChI Key: KEQGOXOOPXJVFM-UHFFFAOYSA-N Synonym: 2-3-dimethylamino propoxy phenyl methanol,2-3-dimethylamino propoxy phenyl,benzenemethanol,2-3-dimethylamino propoxy,2-3-dimethylamino propoxy phenyl methan-1-ol PubChem CID: 23401802 IUPAC Name: [2-[3-(dimethylamino)propoxy]phenyl]methanol SMILES: CN(C)CCCOC1=CC=CC=C1CO