accessibility menu, dialog, popup

UserName

CAS RN 145335-90-6

IUPAC Name: imidazo[1,2-a]pyridine-8-carboxylic acid hydrochloride
Synonyms: imidazo[1,2-a]pyridine-8-carboxylic acid hydrochloride
Molecular Weight (g/mol): 198.61
Molecular Formula: C8H7ClN2O2
InChi Key: XRQZBGADESKIBB-UHFFFAOYSA-N
SMILES: Cl.OC(=O)C1=CC=CN2C=CN=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)