CAS RN 14402-67-6
IUPAC Name:
dipotassium oxotitaniumbis(ylium) dioxalate
Synonyms:
dipotassium oxotitaniumbis(ylium) dioxalate
Molecular Weight (g/mol):
318.1
Molecular Formula:
C4K2O9Ti
InChi Key:
UHWHMHPXHWHWPX-UHFFFAOYSA-J
SMILES:
[K+].[K+].O=[Ti++].[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O
1
–
1
of
1
results
1
Potassium bis(oxalato)oxotitanate(IV) dihydrate
CAS: 14402-67-6 Molecular Formula: C4K2O9Ti Molecular Weight (g/mol): 318.10 MDL Number: MFCD00150592 InChI Key: UHWHMHPXHWHWPX-UHFFFAOYSA-J Synonym: Titanium potassium oxalate IUPAC Name: dipotassium oxotitaniumbis(ylium) dioxalate SMILES: [K+].[K+].O=[Ti++].[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O