accessibility menu, dialog, popup

UserName

CH3H3COH3CH3C

CAS RN 14376-79-5

IUPAC Name: 3,3,5,5-tetramethylcyclohexan-1-one
Synonyms: 3,3,5,5-tetramethylcyclohexan-1-one
Molecular Weight (g/mol): 154.25
Molecular Formula: C10H18O
InChi Key: OQJMHUOCLRCSED-UHFFFAOYSA-N
SMILES: CC1(C)CC(=O)CC(C)(C)C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)