accessibility menu, dialog, popup

UserName

CAS RN 14338-36-4

IUPAC Name: 2-(3-aminophenyl)acetic acid
Synonyms: (3-aminophenyl)acetic acid
Molecular Weight (g/mol): 151.17
Molecular Formula: C8H9NO2
InChi Key: XUSKZLBLGHBCLD-UHFFFAOYSA-N
SMILES: NC1=CC=CC(CC(O)=O)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)