accessibility menu, dialog, popup

UserName

H3CNH3COONCH3CH3

CAS RN 14287-98-0

IUPAC Name: N,N,N',N'-tetrabutylpropanediamide
Synonyms: N,N,N',N'-tetrabutylpropanediamide
Molecular Weight (g/mol): 326.53
Molecular Formula: C19H38N2O2
InChi Key: QTVRIQFMPJRJAK-UHFFFAOYSA-N
SMILES: CCCCN(CCCC)C(=O)CC(=O)N(CCCC)CCCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results