accessibility menu, dialog, popup

UserName

CH3SnCH3H3CSnH3CCH3H3C(E/Z)

CAS RN 14275-61-7

IUPAC Name: tributyl[2-(tributylstannyl)ethenyl]stannane
Synonyms: tributyl[2-(tributylstannyl)ethenyl]stannane
Molecular Weight (g/mol): 606.15
Molecular Formula: C26H56Sn2
InChi Key: VNKOWRBFAJTPLS-UHFFFAOYSA-N
SMILES: CCCC[Sn](CCCC)(CCCC)C=C[Sn](CCCC)(CCCC)CCCC
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)