accessibility menu, dialog, popup

UserName

CAS RN 14268-66-7

IUPAC Name: 2H-1,3-benzodioxol-5-amine
Synonyms: 2H-1,3-benzodioxol-5-amine
Molecular Weight (g/mol): 137.14
Molecular Formula: C7H7NO2
InChi Key: XGNXYCFREOZBOL-UHFFFAOYSA-N
SMILES: NC1=CC=C2OCOC2=C1
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)