accessibility menu, dialog, popup

UserName

CAS RN 142547-16-8

H3COO(R)(R)H3N

CAS RN 142547-16-8

IUPAC Name: (1R,6R)-6-(ethoxycarbonyl)cyclohex-3-en-1-aminium
Synonyms: (1R,6R)-6-(ethoxycarbonyl)cyclohex-3-en-1-aminium
Molecular Weight (g/mol): 170.23
Molecular Formula: C9H16NO2
InChi Key: PTXUIEXYXGXMEU-HTQZYQBOSA-O
SMILES: CCOC(=O)[C@@H]1CC=CC[C@H]1[NH3+]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results