accessibility menu, dialog, popup

UserName

CAS RN 142507-74-2

IUPAC Name: {[2,5-dihydroxy-3,4,6-tris(phosphonooxy)cyclohexyl]oxy}phosphonic acid amine
Synonyms: [2,5-dihydroxy-3,4,6-tris(phosphonooxy)cyclohexyl]oxyphosphonic acid amine
Molecular Weight (g/mol): 517.1
Molecular Formula: C6H19NO18P4
InChi Key: ANQVRWQIKKBLLK-UHFFFAOYNA-N
SMILES: N.OC1C(OP(O)(O)=O)C(OP(O)(O)=O)C(O)C(OP(O)(O)=O)C1OP(O)(O)=O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results