accessibility menu, dialog, popup

UserName

CAS RN 14202-14-3

IUPAC Name: 2-(3,5-dimethyladamantan-1-yl)acetic acid
Synonyms: (3,5-dimethyladamantan-1-yl)acetic acid
Molecular Weight (g/mol): 222.33
Molecular Formula: C14H22O2
InChi Key: FUOXJVUIQUYDDI-UHFFFAOYNA-N
SMILES: CC12CC3CC(C)(C1)CC(CC(O)=O)(C3)C2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results