accessibility menu, dialog, popup

UserName

CH3OCH3CH3OH3C

CAS RN 141-72-0

IUPAC Name: 1-[(2-butoxypropan-2-yl)oxy]butane
Synonyms: 1-[(2-butoxypropan-2-yl)oxy]butane
Molecular Weight (g/mol): 188.31
Molecular Formula: C11H24O2
InChi Key: CXBFTMAIVDLZLG-UHFFFAOYSA-N
SMILES: CCCCOC(C)(C)OCCCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results