CAS RN 1408074-51-0
IUPAC Name:
1-oxa-6-azaspiro[3.4]octane; oxalic acid
Synonyms:
1-oxa-6-azaspiro[3.4]octane; oxalic acid
Molecular Weight (g/mol):
203.19
Molecular Formula:
C8H13NO5
InChi Key:
ZKTJFOHJBNDMDS-UHFFFAOYNA-N
SMILES:
OC(=O)C(O)=O.C1CC2(CCNC2)O1
1
–
1
of
1
results
1
Promotions Available
1-Oxa-6-azaspiro[3.4]octane oxalate, 96%, Thermo Scientific™
1-Oxa-6-azaspiro[3.4]octane oxalate, 96%, Thermo Scientific™
CAS: 1408074-51-0 Molecular Formula: C8H13NO5 Molecular Weight (g/mol): 203.19 MDL Number: MFCD19982774 InChI Key: ZKTJFOHJBNDMDS-UHFFFAOYNA-N Synonym: 1-oxa-6-azaspiro 3.4 octane oxalate,1-oxa-6-azaspiro 3.4 octane; oxalic acid PubChem CID: 71432930 IUPAC Name: 1-oxa-7-azaspiro[3.4]octane;oxalic acid SMILES: OC(=O)C(O)=O.C1CC2(CCNC2)O1