CAS RN 14019-62-6
IUPAC Name:
propan-2-yl 2-aminoacetate hydrochloride
Synonyms:
isopropyl 2-aminoacetate hydrochloride
Molecular Weight (g/mol):
153.61
Molecular Formula:
C5H12ClNO2
InChi Key:
UBKCIXXGQRZHRO-UHFFFAOYSA-N
SMILES:
Cl.CC(C)OC(=O)CN
1
–
1
of
1
results
1
Promotions Available
Glycine isopropyl ester hydrochloride, 96%
Glycine isopropyl ester hydrochloride, 96%
CAS: 14019-62-6 Molecular Formula: C5H12ClNO2 Molecular Weight (g/mol): 153.606 MDL Number: MFCD00151826 InChI Key: UBKCIXXGQRZHRO-UHFFFAOYSA-N Synonym: isopropyl 2-aminoacetate hydrochloride,glycine isopropyl ester hydrochloride,propan-2-yl 2-aminoacetate hydrochloride,glycine, 1-methylethyl ester, hydrochloride,h-gly-oipr?cl,glycine, 1-methylethylester, hydrochloride 1:1 PubChem CID: 23033482 IUPAC Name: propan-2-yl 2-aminoacetate;hydrochloride SMILES: CC(C)OC(=O)CN.Cl