CAS RN 140-92-1
IUPAC Name:
potassium [(propan-2-yloxy)methanethioyl]sulfanide
Synonyms:
proxan potassium
Molecular Weight (g/mol):
174.32
Molecular Formula:
C4H7KOS2
InChi Key:
ZMWBGRXFDPJFGC-UHFFFAOYSA-M
SMILES:
[K+].CC(C)OC([S-])=S
1
–
1
of
1
results
1
Potassium Isopropylxanthate 98.0+%, TCI America™
CAS: 140-92-1 Molecular Formula: C4H7KOS2 Molecular Weight (g/mol): 174.317 MDL Number: MFCD00004930 InChI Key: ZMWBGRXFDPJFGC-UHFFFAOYSA-M Synonym: Isopropylxanthic Acid Potassium Salt PubChem CID: 10931938 IUPAC Name: potassium;propan-2-yloxymethanedithioate SMILES: CC(C)OC(=S)[S-].[K+]