accessibility menu, dialog, popup

UserName

CAS RN 140-64-7

IUPAC Name: bis(2-hydroxyethane-1-sulfonic acid); 4-{[5-(4-carbamimidoylphenoxy)pentyl]oxy}benzene-1-carboximidamide
Synonyms: bis(isethionic acid); pentamidine
Molecular Weight (g/mol): 592.68
Molecular Formula: C23H36N4O10S2
InChi Key: YBVNFKZSMZGRAD-UHFFFAOYSA-N
SMILES: OCCS(O)(=O)=O.OCCS(O)(=O)=O.NC(=N)C1=CC=C(OCCCCCOC2=CC=C(C=C2)C(N)=N)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results