CAS RN 140-40-9
IUPAC Name:
N-(5-nitro-1,3-thiazol-2-yl)acetamide
Synonyms:
2-acetamido-5-nitrothiazole
Molecular Weight (g/mol):
187.17
Molecular Formula:
C5H5N3O3S
InChi Key:
UJRRDDHEMZLWFI-UHFFFAOYSA-N
SMILES:
CC(=O)NC1=NC=C(S1)[N+]([O-])=O
1
–
1
of
1
results
1
2-Acetamido-5-nitrothiazole 98.0+%, TCI America™
CAS: 140-40-9 Molecular Formula: C5H5N3O3S Molecular Weight (g/mol): 187.17 MDL Number: MFCD00022438 InChI Key: UJRRDDHEMZLWFI-UHFFFAOYSA-N Synonym: nithiamide,2-acetamido-5-nitrothiazole,aminitrozole,acinitrazole,nitazole,tritheon,gynofon,ametoterina,aminitrozol,lavoflagin PubChem CID: 8798 IUPAC Name: N-(5-nitro-1,3-thiazol-2-yl)acetamide SMILES: CC(=O)NC1=NC=C(S1)[N+]([O-])=O