accessibility menu, dialog, popup

UserName

OOOO

CAS RN 140-24-9

IUPAC Name: 1,10-dibenzyl decanedioate
Synonyms: 1,10-dibenzyl decanedioate
Molecular Weight (g/mol): 382.5
Molecular Formula: C24H30O4
InChi Key: DVLXEVMGHXWBOZ-UHFFFAOYSA-N
SMILES: O=C(CCCCCCCCC(=O)OCC1=CC=CC=C1)OCC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results