accessibility menu, dialog, popup

UserName

Enjoy Up to 50% Off Spotlight Deals All Year Long

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Main Menu
Documents and Certificates

CAS RN 139896-65-4

H3CSiCH3CH3SSSiCH3CH3CH3

CAS RN 139896-65-4

IUPAC Name: trimethyl(2-{5-[2-(trimethylsilyl)ethynyl]thieno[3,2-b]thiophen-2-yl}ethynyl)silane
Synonyms: trimethyl(2-{5-[2-(trimethylsilyl)ethynyl]thieno[3,2-b]thiophen-2-yl}ethynyl)silane
Molecular Weight (g/mol): 332.63
Molecular Formula: C16H20S2Si2
InChi Key: OWXZBWIZKNUJOL-UHFFFAOYSA-N
SMILES: C[Si](C)(C)C#CC1=CC2=C(S1)C=C(S2)C#C[Si](C)(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.