accessibility menu, dialog, popup

UserName

CAS RN 1393330-40-9

HCOOOOOOONOO

CAS RN 1393330-40-9

IUPAC Name: 2,5-dioxopyrrolidin-1-yl 4,7,10,13,16-pentaoxanonadec-18-ynoate
Synonyms: 2,5-dioxopyrrolidin-1-yl 4,7,10,13,16-pentaoxanonadec-18-ynoate
Molecular Weight (g/mol): 401.41
Molecular Formula: C18H27NO9
InChi Key: ACZDUKRWTPZVRP-UHFFFAOYSA-N
SMILES: O=C(CCOCCOCCOCCOCCOCC#C)ON1C(=O)CCC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results