accessibility menu, dialog, popup

UserName

CAS RN 139332-66-4

NHNNONNOO

CAS RN 139332-66-4

IUPAC Name: 4-nitro-7-(piperazin-1-yl)-2,1,3-benzoxadiazole
Synonyms: 4-nitro-7-(piperazin-1-yl)-2,1,3-benzoxadiazole
Molecular Weight (g/mol): 249.23
Molecular Formula: C10H11N5O3
InChi Key: MVLWYDGJBGPXOL-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(N2CCNCC2)C2=NON=C12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results