accessibility menu, dialog, popup

UserName

OHOHO

CAS RN 139-85-5

IUPAC Name: 3,4-dihydroxybenzaldehyde
Synonyms: 3,4-dihydroxybenzaldehyde protocatechualdehyde rancinamycin IV protocatechuicaldehyde protocatechuic aldehyde
Molecular Weight (g/mol): 138.12
Molecular Formula: C7H6O3
InChi Key: IBGBGRVKPALMCQ-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=O)C=C1O
Molecular Weight (g/mol) 
  • (2)
  • (5)
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (2)
  • (3)
  • (1)
  • (1)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (4)