accessibility menu, dialog, popup

UserName

CAS RN 138891-01-7

IUPAC Name: 3-acetyl-N-methylthiophene-2-sulfonamide
Synonyms: 3-acetyl-N-methylthiophene-2-sulfonamide
Molecular Weight (g/mol): 219.27
Molecular Formula: C7H9NO3S2
InChi Key: OTWLSKTWKAMLMQ-UHFFFAOYSA-N
SMILES: CNS(=O)(=O)C1=C(C=CS1)C(C)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results