accessibility menu, dialog, popup

UserName

CAS RN 13820-41-2

IUPAC Name: platinum(2+) diammonium tetrachloride
Synonyms: platinum(2+) diammonium tetrachloride
Molecular Weight (g/mol): 372.96
Molecular Formula: Cl4H8N2Pt
InChi Key: QJIMNDWDOXTTBR-UHFFFAOYSA-L
SMILES: [NH4+].[NH4+].[Cl-].[Cl-].[Cl-].[Cl-].[Pt++]
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results