accessibility menu, dialog, popup

UserName

CAS RN 13816-33-6

IUPAC Name: 4-(propan-2-yl)benzonitrile
Synonyms: cuminyl nitrile
Molecular Weight (g/mol): 145.21
Molecular Formula: C10H11N
InChi Key: YFDJCWXBKWRDPW-UHFFFAOYSA-N
SMILES: CC(C)C1=CC=C(C=C1)C#N
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)