CAS RN 138116-34-4
IUPAC Name:
(4-aminopyridin-3-yl)methanol
Synonyms:
(4-aminopyridin-3-yl)methanol
Molecular Weight (g/mol):
124.14
Molecular Formula:
C6H8N2O
InChi Key:
WNBVEYMTVDMSFZ-UHFFFAOYSA-N
SMILES:
NC1=CC=NC=C1CO
1
–
1
of
1
results
1
4-Amino-3-pyridinemethanol, 95%
CAS: 138116-34-4 Molecular Formula: C6H8N2O Molecular Weight (g/mol): 124.143 MDL Number: MFCD06203066 InChI Key: WNBVEYMTVDMSFZ-UHFFFAOYSA-N Synonym: 4-aminopyridin-3-yl methanol,4-amino-pyridin-3-yl-methanol,4-aminopyridine-3-methanol,4-amino-3-pyridinemethanol,3-pyridinemethanol, 4-amino,4-amino-3-hydroxymethylpyridine,4-amino-3-pyridyl methan-1-ol,4-amino-3-pyridinyl methanol,rarechem al bd 1386,acmc-1c1ii PubChem CID: 10796739 IUPAC Name: (4-aminopyridin-3-yl)methanol SMILES: C1=CN=CC(=C1N)CO