accessibility menu, dialog, popup

UserName

OSOOOOSOOOOSOOOGaGa

CAS RN 13780-42-2

IUPAC Name: digallium(3+) trisulfate
Synonyms: digallium(3+) trisulfate
Molecular Weight (g/mol): 427.61
Molecular Formula: Ga2O12S3
InChi Key: UXLRWZBQRAWBQA-UHFFFAOYSA-H
SMILES: [Ga+3].[Ga+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Grade 
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)