accessibility menu, dialog, popup

UserName

CAS RN 13709-46-1

IUPAC Name: praseodymium(3+) trifluoride
Synonyms: praseodymium(3+) trifluoride
Molecular Weight (g/mol): 197.9
Molecular Formula: F3Pr
InChi Key: BOTHRHRVFIZTGG-UHFFFAOYSA-K
SMILES: [F-].[F-].[F-].[Pr+3]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results