accessibility menu, dialog, popup

UserName

CAS RN 137-44-0

IUPAC Name: 4,5-dihydro-1H-pyrazol-5-one
Synonyms: 2,4-dihydropyrazol-3-one
Molecular Weight (g/mol): 84.08
Molecular Formula: C3H4N2O
InChi Key: ZRHUHDUEXWHZMA-UHFFFAOYSA-N
SMILES: O=C1CC=NN1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results