CAS RN 137-30-4
CAS RN 137-30-4
IUPAC Name:
zinc(2+) bis((dimethylcarbamothioyl)sulfanide)
Synonyms:
ziram
Molecular Weight (g/mol):
305.8
Molecular Formula:
C6H12N2S4Zn
InChi Key:
DUBNHZYBDBBJHD-UHFFFAOYSA-L
SMILES:
[Zn++].CN(C)C([S-])=S.CN(C)C([S-])=S
1
–
1
of
1
results
1
Zinc Dimethyldithiocarbamate 97.0+%, TCI America™
CAS: 137-30-4 Molecular Formula: C6H12N2S4Zn Molecular Weight (g/mol): 305.796 MDL Number: MFCD00064797 InChI Key: DUBNHZYBDBBJHD-UHFFFAOYSA-L Synonym: ziram,zinc dimethyldithiocarbamate,carbazinc,fuclasin,methazate,zarlate,aazira,cymate,tsimat,zinc bis dimethyldithiocarbamate PubChem CID: 8722 ChEBI: CHEBI:79736 IUPAC Name: zinc;N,N-dimethylcarbamodithioate SMILES: CN(C)C(=S)[S-].CN(C)C(=S)[S-].[Zn+2]