man

Test

2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Main Menu
Documents and Certificates

CAS RN 136470-78-5

NH2NHNNNHON

CAS RN 136470-78-5

IUPAC Name: [(1S,4R)-4-[2-amino-6-(cyclopropylamino)-9H-purin-9-yl]cyclopent-2-en-1-yl]methanol
Synonyms: abacavir ziagen
Molecular Weight (g/mol): 286.34
Molecular Formula: C14H18N6O
InChi Key: MCGSCOLBFJQGHM-SCZZXKLOSA-N
SMILES: NC1=NC(NC2CC2)=C2N=CN([C@@H]3C[C@H](CO)C=C3)C2=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results