accessibility menu, dialog, popup

UserName

CAS RN 13641-96-8

H2COONCO

CAS RN 13641-96-8

IUPAC Name: 2-isocyanatoethyl prop-2-enoate
Synonyms: 2-isocyanatoethyl prop-2-enoate
Molecular Weight (g/mol): 141.13
Molecular Formula: C6H7NO3
InChi Key: DPNXHTDWGGVXID-UHFFFAOYSA-N
SMILES: C=CC(=O)OCCN=C=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results