CAS RN 13617-40-8
CAS RN 13617-40-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Trichloro(3-phenylpropyl)silane 97.0+%, TCI America™
CAS: 13617-40-8 Molecular Formula: C9H11Cl3Si Molecular Weight (g/mol): 253.622 MDL Number: MFCD09953886 InChI Key: QJOHXSVBLYVERP-UHFFFAOYSA-N Synonym: (3-Phenylpropyl)trichlorosilane PubChem CID: 12441988 IUPAC Name: trichloro(3-phenylpropyl)silane SMILES: C1=CC=C(C=C1)CCC[Si](Cl)(Cl)Cl