CAS RN 13602-12-5
IUPAC Name:
4-carboxypyridin-1-ium-1-olate
Synonyms:
isonicotinic acid N-oxide
Molecular Weight (g/mol):
139.11
Molecular Formula:
C6H5NO3
InChi Key:
QCWTWMJMLSKQCJ-UHFFFAOYSA-N
SMILES:
OC(=O)C1=CC=[N+]([O-])C=C1
1
–
1
of
1
results
1
Isonicotinic Acid N-Oxide 98.0+%, TCI America™
CAS: 13602-12-5 Molecular Formula: C6H5NO3 Molecular Weight (g/mol): 139.11 MDL Number: MFCD00006209 InChI Key: QCWTWMJMLSKQCJ-UHFFFAOYSA-N Synonym: isonicotinic acid n-oxide,4-carboxypyridine 1-oxide,pyridine-4-carboxylic acid n-oxide,4-pyridinecarboxylic acid 1-oxide,4-pyridinecarboxylic acid, 1-oxide,pyridine-4-carboxylic acid 1-oxide,4-carboxypyridin-1-ium-1-olate,isonicotinic acid 1-oxide,1-hydroxypyridine-4-carboxylic acid,pyridin-n-oxide-4-carboxylic acid PubChem CID: 26130 IUPAC Name: 4-carboxypyridin-1-ium-1-olate SMILES: OC(=O)C1=CC=[N+]([O-])C=C1