accessibility menu, dialog, popup

UserName

HClNH2NNH2NN(E/Z)

CAS RN 136-40-3

IUPAC Name: hydrogen 3-(2-phenyldiazen-1-yl)pyridine-2,6-diamine chloride
Synonyms: hydrogen phenazopyridine chloride
Molecular Weight (g/mol): 249.7
Molecular Formula: C11H12ClN5
InChi Key: QQBPIHBUCMDKFG-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NC1=CC=C(N=NC2=CC=CC=C2)C(N)=N1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)