accessibility menu, dialog, popup

UserName

OOOOZnW

CAS RN 13597-56-3

IUPAC Name: zinc(2+) tungsten tetraoxidandiide
Synonyms: zinc(2+) tungsten tetraoxidandiide
Molecular Weight (g/mol): 313.22
Molecular Formula: O4WZn
InChi Key: IGKCQWHBCHMDFW-UHFFFAOYSA-N
SMILES: [O--].[O--].[O--].[O--].[Zn++].[W]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results