accessibility menu, dialog, popup

UserName

NaNaNaNaOSiOOO

CAS RN 13472-30-5

IUPAC Name: tetrasodium silicate
Synonyms: sodium orthosilicate
Molecular Weight (g/mol): 184.04
Molecular Formula: Na4O4Si
InChi Key: POWFTOSLLWLEBN-UHFFFAOYSA-N
SMILES: [Na+].[Na+].[Na+].[Na+].[O-][Si]([O-])([O-])[O-]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results