accessibility menu, dialog, popup

UserName

CAS RN 13465-77-5

ClSiClClSiClClCl

CAS RN 13465-77-5

IUPAC Name: hexachlorodisilane
Synonyms: hexachlorodisilane
Molecular Weight (g/mol): 268.87
Molecular Formula: Cl6Si2
InChi Key: LXEXBJXDGVGRAR-UHFFFAOYSA-N
SMILES: Cl[Si](Cl)(Cl)[Si](Cl)(Cl)Cl
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results