CAS RN 13465-10-6
IUPAC Name:
λ¹-indiganyliumylidene chloride
Synonyms:
λ¹-indiganyliumylidene chloride
Molecular Weight (g/mol):
150.27
Molecular Formula:
ClIn
InChi Key:
APHGZSBLRQFRCA-UHFFFAOYSA-M
SMILES:
[Cl-].[In+]
1
–
1
of
1
results
1
Indium(I) chloride, Puratronic™, 99.999% (metals basis)
CAS: 13465-10-6 Molecular Formula: ClIn Molecular Weight (g/mol): 150.27 MDL Number: MFCD00148875 InChI Key: APHGZSBLRQFRCA-UHFFFAOYSA-M IUPAC Name: λ¹-indiganyliumylidene chloride SMILES: [Cl-].[In+]