accessibility menu, dialog, popup

UserName

CAS RN 13450-92-5

IUPAC Name:
Synonyms:
Molecular Weight (g/mol): 400.31
Molecular Formula: Br4GeH8
InChi Key: RLMRZMHUNFSBLI-UHFFFAOYSA-N
SMILES: [GeH4].Br.Br.Br.Br
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results