accessibility menu, dialog, popup

UserName

CAS RN 13446-24-7

MgMgOPOOOPOOO

CAS RN 13446-24-7

IUPAC Name: dimagnesium(2+) (phosphonatooxy)phosphonate
Synonyms: dimagnesium(2+) diphosphate
Molecular Weight (g/mol): 222.55
Molecular Formula: Mg2O7P2
InChi Key: XZTWHWHGBBCSMX-UHFFFAOYSA-J
SMILES: [Mg++].[Mg++].[O-]P([O-])(=O)OP([O-])([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results