accessibility menu, dialog, popup

UserName

MnH3CCH3OOH3CCH3OO

CAS RN 13434-24-7

IUPAC Name: manganese(2+) bis(2-ethylhexanoate)
Synonyms: manganese(2+) bis(2-ethylhexanoate)
Molecular Weight (g/mol): 341.35
Molecular Formula: C16H30MnO4
InChi Key: FHRAKXJVEOBCBQ-UHFFFAOYNA-L
SMILES: [Mn++].CCCCC(CC)C([O-])=O.CCCCC(CC)C([O-])=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results