accessibility menu, dialog, popup

UserName

H3COOCH2

CAS RN 13402-02-3

IUPAC Name: hexadecyl prop-2-enoate
Synonyms: hexadecyl prop-2-enoate
Molecular Weight (g/mol): 296.5
Molecular Formula: C19H36O2
InChi Key: PZDUWXKXFAIFOR-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCOC(=O)C=C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results