CAS RN 1337532-51-0
IUPAC Name:
5-bromo-7-methyl-7H-pyrrolo[2,3-d]pyrimidin-4-amine
Synonyms:
5-bromo-7-methylpyrrolo[2,3-d]pyrimidin-4-amine
Molecular Weight (g/mol):
227.07
Molecular Formula:
C7H7BrN4
InChi Key:
SCHJLAFNBDGWJN-UHFFFAOYSA-N
SMILES:
CN1C=C(Br)C2=C(N)N=CN=C12
1
–
1
of
1
results
1
Promotions Available
6-Amino-7-bromo-9-methyl-7-deazapurine, 97%, Thermo Scientific Chemicals
6-Amino-7-bromo-9-methyl-7-deazapurine, 97%, Thermo Scientific Chemicals
CAS: 1337532-51-0 Molecular Formula: C7H7BrN4 Molecular Weight (g/mol): 227.065 MDL Number: MFCD20486185 InChI Key: SCHJLAFNBDGWJN-UHFFFAOYSA-N Synonym: 5-bromo-7-methyl-7h-pyrrolo 2,3-d pyrimidin-4-amine,5-bromo-7-methylpyrrolo 2,3-d pyrimidin-4-amine,6-amino-7-bromo-9-methyl-7-deazapurine,5-bromo-7-methyl-7h-pyrrolo-2,3-d pyrimidin-4-amine,7h-pyrrolo 2,3-d pyrimidin-4-amine,5-bromo-7-methyl,7h-pyrrolo 2,3-d pyrimidin-4-amine, 5-bromo-7-methyl PubChem CID: 66560938 IUPAC Name: 5-bromo-7-methylpyrrolo[2,3-d]pyrimidin-4-amine SMILES: CN1C=C(C2=C1N=CN=C2N)Br